missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 11447597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447597 1 g 1g
11457597 5 g 5g
11467597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447597 Supplier Thermo Scientific Alfa Aesar Supplier No. A15667.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346-06-5
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00004621
InChI Key TWQNSHZTQSLJEE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzyl alcohol, 2-trifluoromethyl phenyl methanol, o-trifluoromethylbenzyl alcohol, 2-trifluoromethyl benzylalcohol, 2-trifluoromethylbenzyl alcohol, o-trifluoromethyl benzyl alcohol, 2-trifluoromethyl benzylic alcohol, benzenemethanol, 2-trifluoromethyl, 2-trifluoromethyl phenyl methan-1-ol
PubChem CID 67666
IUPAC Name [2-(trifluoromethyl)phenyl]methanol
SMILES OCC1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 4°C
Density 1.326
Boiling Point 80°C to 83°C (3 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.468
Quantity 1 g
Beilstein 2093714
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.