missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)thiophenol, 97%

Product Code. 11329945
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329945 1 g 1g
11339945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329945 Supplier Thermo Scientific Alfa Aesar Supplier No. L19911.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 175278-01-0 | C7H5F3OS | 194.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-01-0
Molecular Formula C7H5F3OS
Molecular Weight (g/mol) 194.17
MDL Number MFCD00052320
InChI Key CIIRSPLZNQRPGX-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy thiophenol, 2-trifluoromethoxy benzenethiol, 2-trifluoromethoxy benzene-1-thiol, benzenethiol, 2-trifluoromethoxy, acmc-1ca0h, 2-trifluoromethoxythiophenol, 2-trifluoromethoxy-thiophenol, 2-trifluoromethoxy-benzenethiol, 2-trifluoromethyloxy benzenethiol
PubChem CID 2760732
SMILES FC(F)(F)OC1=CC=CC=C1S

Specifications

Boiling Point 53°C to 55°C (15 mmHg)
Refractive Index 1.4768
Quantity 1 g
UN Number UN2810
Sensitivity Air Sensitive
IUPAC Name 2-(trifluoromethoxy)benzenethiol
Formula Weight 194.18
Percent Purity 99%
Chemical Name or Material 2-(Trifluoromethoxy)thiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.