missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophene Carboxaldehyde, 99.92%, MP Biomedicals™

Product Code. 11405301 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405301 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405301 Supplier MP Biomedicals Supplier No. 0215688725

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 98-03-3
Color Yellow
Molecular Formula C5H4OS
MDL Number MFCD00005429
Quantity 25 g
Synonym 2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
InChI Key CNUDBTRUORMMPA-UHFFFAOYSA-N
SMILES O=CC1=CC=CS1
IUPAC Name thiophene-2-carbaldehyde
Molecular Weight (g/mol) 112.15
PubChem CID 7364
ChEBI CHEBI:87301
Formula Weight 112.1
Physical Form Liquid
Chemical Name or Material 2-Thiophene Carboxaldehyde, FTIR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.