missing translation for 'onlineSavingsMsg'
Learn More

2-Tetradecanol, TRC

Product Code. 30417872
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417872 2.5 g 2.5g
30426297 250 mg 250mg
30414217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417872 Supplier Toronto Research Chemicals Supplier No. TRCT2045152.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4706-81-4
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.39
Synonym (±)-2-Tetradecanol, 1-Methyltridecyl Alcohol, NSC 87599
IUPAC Name tetradecan-2-ol
SMILES CCCCCCCCCCCCC(C)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H30O/c1-3-4-5-6-7-8-9-10-11-12-13-14(2)15/h14-15H,3-13H2,1-2H3
Formula Weight 214.2297
Chemical Name or Material 2-Tetradecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.