accessibility menu, dialog, popup

UserName

CAS RN 4706-81-4

H3CCH3OH

CAS RN 4706-81-4

IUPAC Name: tetradecan-2-ol
Synonyms: 2-tetradecanol
Molecular Weight (g/mol): 214.39
Molecular Formula: C14H30O
InChi Key: BRGJIIMZXMWMCC-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCC(C)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results