Learn More
2-sec-Butyl-d5-4-{4-[4-(4-hydroxy-phenyl)-piperazin-1-yl]-phenyl}-2,4-dihydro-[1,2,4]-triazol-3-one, TRC
Product Code. 30340850
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340850 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30340850 Supplier Toronto Research Chemicals Supplier No. TRCB69202750MG
 Restricted Item
Estimated Shipment: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-sec-Butyl-d5-4-{4-[4-(4-hydroxy-phenyl)-piperazin-1-yl]-ph
CAS 1020719-21-4
Molecular Formula C22 2H5 H22 N5 O2
Quantity 50 mg
InChI Formula InChI=1S/C22H27N5O2/c1-3-17(2)27-22(29)26(16-23-27)20-6-4-18(5-7-20)24-12-14-25(15-13-24)19-8-10-21(28)11-9-19/h4-11,16-17,28H,3,12-15H2,1-2H3/i1D3,3D2
SMILES [2H]C([2H])([2H])C([2H])([2H])C(C)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(O)cc4
IUPAC Name 4-[4-[4-(4-hydroxyphenyl)piperazin-1-yl]phenyl]-2-(3,3,4,4,4-pentadeuteriobutan-2-yl)-1,2,4-triazol-3-one
Molecular Weight (g/mol) 398.51
Formula Weight 398.2479