Learn More
2-Pyridineacetonitrile, 97%
Product Code. p-7020955
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424414 5 g 5g
11434414 25 g 25g
11444414 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11424414 Supplier Thermo Scientific Alfa Aesar Supplier No. A13087.06
Request Bulk or Custom Format
CAS: 2739-97-1 | C7H6N2 | 118.139 g/mol
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2739-97-1
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00006346
InChI Key UKVQBONVSSLJBB-UHFFFAOYSA-N
Synonym 2-pyridylacetonitrile, 2-pyridineacetonitrile, pyridine-2-acetonitrile, 2-pyridin-2-yl acetonitrile, 2-2-pyridyl acetonitrile, 2-cyanomethyl pyridine, pyridine-acetonitrile, pyridyl-2-acetonitrile, pyridin-2-ylacetonitrile, pyridin-2-yl acetonitrile
PubChem CID 75959
IUPAC Name 2-pyridin-2-ylacetonitrile
SMILES C1=CC=NC(=C1)CC#N
Specifications
Melting Point 23°C to 25°C
Density 1.059
Boiling Point 66°C to 68°C (1 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.525
Quantity 5 g
UN Number UN3439
Beilstein 111215
Solubility Information Miscible with water.
Formula Weight 118.14
Percent Purity 99%
Chemical Name or Material 2-Pyridineacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • Pyridine-2-acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Pyridine-2-acetonitrile