Learn More
2-(Piperidin-1-yl)ethyl 3-methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylate hydrochloride, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 3717-88-2 |
| Molecular Formula | C24H26ClNO4 |
| MDL Number | MFCD00072099 |
| Quantity | 25 g |
| SMILES | O=C(C1=C2C(C(C(C)=C(C3=CC=CC=C3)O2)=O)=CC=C1)OCCN4CCCCC4.Cl |
| Molecular Weight (g/mol) | 427.92 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-(Piperidin-1-yl)ethyl 3-methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylate hydrochloride |