Learn More
2-(Piperazin-1-yl)pyrimidine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 20980-22-7 |
| Molecular Formula | C8H12N4 |
| MDL Number | MFCD00040742 |
| Quantity | 100 g |
| SMILES | C1(N2CCNCC2)=NC=CC=N1 |
| Molecular Weight (g/mol) | 164.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(Piperazin-1-yl)pyrimidine |