Learn More
2-Oxoindoline-5-sulfonyl chloride, 97%, Thermo Scientific™
Product Code. 10498593
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498593 1 g Amber Glass Bottle 1g
10367584 250 mg Amber Glass Bottle 250mg
10205664 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10498593 Supplier Thermo Scientific Maybridge Supplier No. CC53303DA
 Restricted Item
Estimated Shipment: 18-11-2026
Chemical Identifiers
CAS 199328-31-9
Molecular Formula C8H6ClNO3S
Molecular Weight (g/mol) 231.65
MDL Number MFCD06655924
InChI Key FYBNVKMJOOYPGI-UHFFFAOYSA-N
Synonym 2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole
PubChem CID 4962506
SMILES ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1
Specifications
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride
Formula Weight 231.66
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Oxoindoline-5-sulfonyl chloride