Learn More
2-O-Benzyl-1,3,4,6-tetra-O-acetyl-Alpha-D-mannopyranose, TRC
Specifications
Specifications
| Chemical Name or Material | 2-O-Benzyl-1,3,4,6-tetra-O-acetyl-a-D-mannopyranose |
| CAS | 80779-87-9 |
| Molecular Formula | C21 H26 O10 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C21H26O10/c1-12(22)26-11-17-18(28-13(2)23)19(29-14(3)24)20(21(31-17)30-15(4)25)27-10-16-8-6-5-7-9-16/h5-9,17-21H,10-11H2,1-4H3/t17-,18-,19+,20+,21+/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H](OC(=O)C)[C@@H](OCc2ccccc2)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | [(2R,3R,4S,5S,6R)-3,4,6-triacetyloxy-5-phenylmethoxyoxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 438.43 |
| Formula Weight | 438.1526 |