Learn More
2-Nonyn-1-ol, 96%
Product Code. 11332496
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11332496
Brand: Thermo Scientific Alfa Aesar L06380.14
Request Bulk or Custom Format
CAS: 5921-73-3 | C9H16O | 140.23 g/mol
2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5921-73-3
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00041566
InChI Key XERDOEKKHDALKJ-UHFFFAOYSA-N
Synonym 2-nonyn-1-ol, pubchem13067, acmc-1apv0
PubChem CID 80017
IUPAC Name non-2-yn-1-ol
SMILES CCCCCCC#CCO
Specifications
Density 0.873
Boiling Point 109°C to 111°C (11 mmHg)
Refractive Index 1.456
Quantity 25 g
Beilstein 1745753
Solubility Information Not miscible in water.
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 2-Nonyn-1-ol