Learn More
2’-Nitrophenyl 2,3,4-Tri-O-acetyl-Beta-D-xylopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 2'-Nitrophenyl 2,3,4-Tri-O-acetyl-beta-D-xylopyranoside |
| CAS | 10256-24-3 |
| Molecular Formula | C17 H19 N O10 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C17H19NO10/c1-9(19)25-14-8-24-17(16(27-11(3)21)15(14)26-10(2)20)28-13-7-5-4-6-12(13)18(22)23/h4-7,14-17H,8H2,1-3H3/t14-,15+,16-,17+/m1/s1 |
| SMILES | CC(=O)O[C@@H]1CO[C@@H](Oc2ccccc2[N+](=O)[O-])[C@H](OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | [(3R,4S,5R,6S)-4,5-diacetyloxy-6-(2-nitrophenoxy)oxan-3-yl] acetate |
| Molecular Weight (g/mol) | 397.33 |
| Formula Weight | 397.1009 |