missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrofuran, TRC

Product Code. 30475557
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30475557 250 mg -
30469237 1 g -
30473486 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30475557 Supplier Toronto Research Chemicals Supplier No. TRCN493840250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-39-2
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.07
Synonym NSC 59983, Nitrofuran
IUPAC Name 2-nitrofuran
SMILES [O-][N+](=O)c1occc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C4H3NO3/c6-5(7)4-2-1-3-8-4/h1-3H
Formula Weight 113.0113
Chemical Name or Material 2-Nitrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.