missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Methylvaleraldehyde, 98%
Description
Applications
2-Methylvaleraldehyde is used as an intermediate to manufacture organic compounds (pharmaceuticals, odorants and flavorings)
Solubility
Soluble in water (4.2g/L at 25°C).
2-Methylvaleraldehyde is used as an intermediate to manufacture organic compounds (pharmaceuticals, odorants and flavorings)
Solubility
Soluble in water (4.2g/L at 25°C).
Specifications
Specifications
| CAS | 123-15-9 |
| Melting Point | -100°C |
| Boiling Point | 119°C to 120°C |
| Flash Point | 16°C (62°F) |
| Odor | Like Aldehyde |
| Molecular Formula | C6H12O |
| MDL Number | MFCD00006986 |
| Quantity | 100 g |
| UN Number | UN2367 |
| Beilstein | 1739423 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?