Learn More
2-Methylimidazole, MP Biomedicals
Product Code. 11407285 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11407285 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11407285 Supplier MP Biomedicals Supplier No. 0520938180
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Spezifikation
CAS 693-98-1
Molecular Formula C4H6N2
MDL Number MFCD00005190
Quantity 100 g
Synonym 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
SMILES CC1=NC=CN1
IUPAC Name 2-methyl-1H-imidazole
Molecular Weight (g/mol) 82.11
PubChem CID 12749
Formula Weight 82.11
Chemical Name or Material 2-Methylimidazole
Mehr anzeigen Weniger anzeigen