Learn More
2-Methylbutyrylglycine-d2, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methylbutyrylglycine-d2 |
| CAS | 2469035-10-5 |
| Molecular Formula | C7 D2 H11 N O3 |
| Quantity | 2.5 mg |
| Synonym | N-(2-Methyl-1-oxobutyl)glycine-2,2-d2, N-(2-Methylbutanoyl)glycine-2,2-d2, 2-Methylbutyrylglycine-d2, N-(2-Methylbutanoyl)glycine-2,2-D2 |
| InChI Formula | InChI=1S/C7H13NO3/c1-3-5(2)7(11)8-4-6(9)10/h5H,3-4H2,1-2H3,(H,8,11)(H,9,10)/i4D2 |
| SMILES | [2H]C([2H])(NC(=O)C(C)CC)C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(2-methylbutanoylamino)acetic acid |
| Molecular Weight (g/mol) | 161.195 |
| Formula Weight | 161.102 |