missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, MilliporeSigma™

Product Code. 15804770
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15804770 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15804770 Supplier Supelco Supplier No. 1060561000
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-78-4
Color Colorless
Density 0.62g/cm3 (20°C)
Boiling Point 28°C
Flash Point -57°C (70.6°F)
Odor Characteristic
Packaging Glass Bottle
Molecular Formula C5H12
MDL Number MFCD00009338
Quantity 1 L
UN Number UN 1265
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
Solubility Information Water: 0.048g/L at 48°C (118°F)
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
SMILES CCC(C)C
IUPAC Name 2-methylbutane
Molecular Weight (g/mol) 72.15
PubChem CID 6556
ChEBI CHEBI:30362
Formula Weight 72.15
Vapor Pressure 800 hPa (20°C)
Physical Form Liquid
Chemical Name or Material 2-Methylbutane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.