Learn More
2-Methylbenzenamine Hydrochloride (o-Toluidine Hydrochloride), Mikromol™
Description
2-Methylbenzenamine Hydrochloride (o-Toluidine Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 2-methylbenzenamine hydrochloride |
| CAS | 636-21-5 |
| Analyte or Component Names | 2-methylbenzenamine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H9 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | o-Toluidine Hydrochloride, 2-Methylbenzenamine hydrochloride, Prilocaine Hydrochloride Imp. B (EP), Prilocaine Imp. B (EP), Prilocaine USP Related Compound A, Prilocaine USP RC A, Prilocaine Related Compound A as Hydrochloride, Prilocaine Hydrochloride Impurity B as Hydrochloride, Prilocaine Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C7H9N.ClH/c1-6-4-2-3-5-7(6)8;/h2-5H,8H2,1H3;1H |
| SMILES | Cl.Cc1ccccc1N |
| IUPAC Name | 2-methylaniline;hydrochloride |
| Show More |