missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals

Product Code. p-7038743
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15483686 250 mg 250mg
15493686 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483686 Supplier Thermo Scientific Alfa Aesar Supplier No. H57776.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (1.00E+06 mg/L).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1158098-73-7
Molecular Formula C7H13NO2S
Molecular Weight (g/mol) 175.246
MDL Number MFCD14702522
InChI Key VKUZMNXQGKBLHN-UHFFFAOYSA-N
Synonym 2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide
PubChem CID 46839995
IUPAC Name 2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
SMILES CC(C)(C)S(=O)N=C1COC1

Specifications

Quantity 250 mg
Solubility Information Soluble in water. (1.00E+06mg/L).
Formula Weight 175.25
Percent Purity 95%
Chemical Name or Material 2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.