Learn More
2-Methyl-4-octanone, 99%
Product Code. 11458817
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458817 5 g 5g
11468817 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11458817 Supplier Thermo Scientific Alfa Aesar Supplier No. A19952.06
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
CAS: 7492-38-8 | C9H18O | 142.24 g/mol
2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-4-octanone is a building block ketone for proteomics research. It is also used as pharmaceutical intermediates.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7492-38-8
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00048808
InChI Key ICSKJDZASFIJQK-UHFFFAOYSA-N
Synonym 2-methyl-4-octanone, 4-octanone, 2-methyl
PubChem CID 522566
IUPAC Name 2-methyloctan-4-one
SMILES CCCCC(=O)CC(C)C
Specifications
Boiling Point 180°C
Refractive Index 1.415
Quantity 5 g
Formula Weight 142.24
Percent Purity 99%
Chemical Name or Material 2-Methyl-4-octanone