Learn More
2-Methyl-3-(trifluoromethyl)aniline, 98%, Thermo Scientific™
Artikelnummer. 11400461
Change view
Click to view available options
Quantity:
1 g
5 g
Packungsgröße:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
11400461 1 g 1g
11410461 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11400461 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. B20009.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 54396-44-0
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00153216
InChI Key TWLDBACVSHADLI-UHFFFAOYSA-N
Synonym 2-methyl-3-trifluoromethyl aniline, 3-amino-2-methylbenzotrifluoride, 3-trifluoromethyl-o-toluidine, 3-amino-2-methyl benzotrifluoride, 2-methyl-3-trifluoromethyl-phenylamine, 2-methyl-3 trifluoromethyl aniline, 2-methyl-3-trifluoromethyl benzenamine, benzenamine, 2-methyl-3-trifluoromethyl
PubChem CID 2735932
IUPAC Name 2-methyl-3-(trifluoromethyl)aniline
SMILES CC1=C(C=CC=C1N)C(F)(F)F
Spezifikation
Melting Point 37°C to 40°C
Boiling Point 62°C to 64°C (4 mmHg)
Flash Point 98°C (208°F)
Quantity 1 g
UN Number UN2811
Beilstein 2832865
Formula Weight 175.16
Percent Purity 98%
Chemical Name or Material 2-Methyl-3-(trifluoromethyl)aniline