Learn More
2-Methyl-1-nitronaphthalene, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 881-03-8 |
| Molecular Formula | C11H9NO2 |
| MDL Number | MFCD00003914 |
| Quantity | 10 g |
| SMILES | O=[N+](C1=C2C=CC=CC2=CC=C1C)[O-] |
| Molecular Weight (g/mol) | 187.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Methyl-1-nitronaphthalene |