Learn More
2-Methoxypyridine-5-boronic acid, 95%
Product Code. 11314526
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314526 1 g 1g
11334526 250 mg 250mg
11324526 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11314526 Supplier Thermo Scientific Alfa Aesar Supplier No. L20087.03
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
CAS: 163105-89-3 | C6H8BNO3 | 152.94 g/mol
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 163105-89-3
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD02093044
InChI Key DHADXDMPEUWEAS-UHFFFAOYSA-N
Synonym 2-methoxy-5-pyridineboronic acid, 6-methoxypyridin-3-yl boronic acid, 2-methoxypyridine-5-boronic acid, 2-methoxy-5-pyridinylboronic acid, 6-methoxypyridine-3-boronic acid, 6-methoxypyridin-3-yl-3-boronic acid, 6-methoxy-pyridine-3-boronic acid, 6-methoxy-3-pyridinylboronic acid, 6-methoxy-3-pyridylboronic acid, 6-methoxypyridin-3-yl boranediol
PubChem CID 2734368
IUPAC Name (6-methoxypyridin-3-yl)boronic acid
SMILES COC1=CC=C(C=N1)B(O)O
Spécification
Melting Point 132°C to 133°C
Quantity 1 g
Beilstein 9119950
Solubility Information Soluble in water.
Formula Weight 152.95
Percent Purity 95%
Chemical Name or Material 2-Methoxypyridine-5-boronic acid
RUO – Research Use Only
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.