Learn More
2-Methoxyphenothiazine, Mikromol™
Product Code. 30051299
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051299 100 mg 100mg
30049828 25 mg 25mg
30458601 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30051299 Supplier Mikromol Supplier No. MM0019.02
 Restricted Item
Estimated Shipment: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methoxyphenothiazine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methoxyphenothiazine
CAS 1771-18-2
Analyte or Component Names 2-Methoxyphenothiazine
Shipping Condition Room Temperature
Molecular Formula C13 H11 N O S
Certifications/Compliance ISO 17025
Synonym Levomepromazine Maleate Imp. A (EP), Levomepromazine Imp. A (EP), 2-Methoxyphenothiazine, Levomepromazine Maleate Impurity A, Levomepromazine Impurity A
InChI Formula InChI=1S/C13H11NOS/c1-15-9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)16-13/h2-8,14H,1H3
SMILES COc1ccc2Sc3ccccc3Nc2c1
IUPAC Name 2-methoxy-10H-phenothiazine
Molecular Weight (g/mol) 229.3
Quantity 100 mg
Formula Weight 229.0561 g/mol