missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenol, Sigma-Aldrich™

Product Code. 15860351
Change view
Click to view available options
Quantity:
5 mL
250 mL
Unit Size:
250mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860351 250 mL 250mL
15850351 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15860351 Supplier Sigma Aldrich Supplier No. 8183390250
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-05-1
Grade Research
Molecular Weight (g/mol) 124.14

Specifications

Melting Point 28°C to 32°C
Density 1.13 g/cm3 at 21°C
Boiling Point 205°C/1013 hPa
MDL Number MFCD00002185
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.