missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethoxy)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15427595
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427595 250 mg 250mg
11425960 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427595 Supplier Thermo Scientific Alfa Aesar Supplier No. H26540.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 773109-13-0
Molecular Formula C10H9F3O4
Molecular Weight (g/mol) 250.17
MDL Number MFCD04972913
InChI Key HDDOJDLMPLUGCV-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethoxy phenylacetic acid, 2-2-methoxy-5-trifluoromethoxy phenyl acetic acid, 2-methoxy-5-trifluoromethoxy phenyl acetic acid, 2-methoxy-5-trifluoromethoxyphenyl acetic acid
PubChem CID 17999353
IUPAC Name 2-[2-methoxy-5-(trifluoromethoxy)phenyl]acetic acid
SMILES COC1=C(CC(O)=O)C=C(OC(F)(F)F)C=C1

Specifica

Quantity 250 mg
Formula Weight 250.17
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)phenylacetic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.