Learn More
2-Mercaptothiazoline, TRC
Product Code. 30477243
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477243 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30477243 Supplier Toronto Research Chemicals Supplier No. TRCM225550250G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Mercaptothiazoline
CAS 96-53-7
Molecular Formula C3H5NS2
Quantity 250 g
Synonym 2-Thiazolidinethione, 1,3-Thiazolidine-2-thione, 2-Mercapto-4,5-dihydrothiazole, 2-Thiazoline-2-thiol, 2-Thiothiazolidone, 4,5-Dihydro-2-mercaptothiazole, 4,5-Dihydrothiazole-2-thiol, Metabasal, NSC 680, Sancelent 2MT, Tetrahydrothiazole-2-thione, Thyroidan, WR 305
InChI Formula InChI=1S/C3H5NS2/c5-3-4-1-2-6-3/h1-2H2,(H,4,5)
SMILES S=C1NCCS1
IUPAC Name 1,3-thiazolidine-2-thione
Molecular Weight (g/mol) 119.21
Formula Weight 118.9863