missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopurine, MP Biomedicals™

Product Code. 11497642 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497642 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11497642 Supplier MP Biomedicals Supplier No. 0215535680
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 28128-19-0
Color Orange
Molecular Formula C5H4N4S
MDL Number MFCD00047145
Quantity 100 mg
Synonym 2-mercaptopurine, 2-thiopurine, 5-mercaptopurine, purine-2-thiol, 2-purinethiol, 7h-purine-2-thiol, 9h-purine-2-thiol, 2-mp, 1,3-dihydro-2h-purine-2-thione, 3,7-dihydro-2h-purine-2-thione
InChI Key HDBQZGJWHMCXIL-UHFFFAOYSA-N
SMILES S=C1NC2=C(NC=N2)C=N1
IUPAC Name 3,7-dihydro-2H-purine-2-thione
Molecular Weight (g/mol) 152.18
PubChem CID 3015569
Formula Weight 152.18
Physical Form Powder
Chemical Name or Material 2-Mercaptopurine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.