Learn More
Alfa Aesar™ 2-Mercapto-6-nitrobenzothiazole, 96%
Product Code. 11433747
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433747 5 g 5g
11423747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433747 Supplier Alfa Aesar™ Supplier No. A18211.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 4845-58-3
Molecular Formula C7H4N2O2S2
Molecular Weight (g/mol) 212.24
MDL Number MFCD00041850
InChI Key QPOZGXKWWKLJDK-UHFFFAOYSA-N
Synonym 2-mercapto-6-nitrobenzothiazole, 6-nitrobenzo d thiazole-2-thiol, 6-nitro-2-mercaptobenzothiazole, 6-nitrobenzo d thiazole-2 3h-thione, 6-nitrobenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-nitro, usaf ek-3991, 6-nitro-1,3-benzothiazole-2-thiol, 2-benzothiazolinethione, 6-nitro, 2-benzothiazolethiol, 6-nitro
PubChem CID 947375
IUPAC Name 6-nitro-3H-1,3-benzothiazole-2-thione
SMILES [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1
Specifications
Melting Point 250°C to 254°C
Quantity 5 g
Formula Weight 212.24
Percent Purity 96%
Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole