missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Isopropylamino-1-(2-methoxyphenyl)ethanol

Product Code. 11464660
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464660 1 g 1g
11454660 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464660 Supplier Alfa Aesar™ Supplier No. H50735.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23299-28-7
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.289
MDL Number MFCD08060962
InChI Key RLJUZSQHQRIUSG-UHFFFAOYSA-N
Synonym 2-isopropylamino-1-2-methoxyphenyl ethanol, 2-isopropylamino-1-2-methoxy-phenyl-ethanol, acmc-20ansk, 1-2-methoxyphenyl-2-methylethyl amino ethan-1-ol, 1-2-methoxyphenyl-2-propan-2-yl amino ethan-1-ol, benzenemethanol,2-methoxy-a-1-methylethyl amino methyl
PubChem CID 24208809
IUPAC Name 1-(2-methoxyphenyl)-2-(propan-2-ylamino)ethanol
SMILES CC(C)NCC(C1=CC=CC=C1OC)O

Specifications

Quantity 1 g
Formula Weight 209.29
Chemical Name or Material 2-Isopropylamino-1-(2-methoxyphenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.