missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-6-propylphenol, TRC

Product Code. 30012131
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012131 50 mg 50mg
30003656 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012131 Supplier Toronto Research Chemicals Supplier No. TRCI87289550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74663-48-2
Molecular Formula C12 H18 O
Molecular Weight (g/mol) 178.27
Synonym 2-(1-Methylethyl)-6-propylphenol
IUPAC Name 2-propan-2-yl-6-propylphenol
SMILES CCCc1cccc(C(C)C)c1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H18O/c1-4-6-10-7-5-8-11(9(2)3)12(10)13/h5,7-9,13H,4,6H2,1-3H3
Formula Weight 178.1358
Chemical Name or Material 2-Isopropyl-6-propylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.