missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyphenylboronic acid, 97%, Thermo Scientific™

Product Code. p-4258880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11975881 1 g 1g
11995881 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975881 Supplier Thermo Scientific Acros Supplier No. 431380010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 138008-97-6
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03427050
InChI Key XDMKBIIRBDPSOE-UHFFFAOYSA-N
Synonym 2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297
PubChem CID 5084099
IUPAC Name (2-propan-2-yloxyphenyl)boronic acid
SMILES CC(C)OC1=CC=CC=C1B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 39.0°C to 43.0°C
Color White
Boiling Point 199.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Isopropoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.