missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyethanol, TRC

Product Code. 30008316
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008316 100 mL 100mL
30007394 250 mL 250mL
30004603 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008316 Supplier Toronto Research Chemicals Supplier No. TRCI918253100ML
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-59-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
Synonym 2-(1-Methylethoxy)ethanol, 2-(Propan-2-yloxy)ethanol, 2-Isopropyloxyethanol, 2-[(1-Methylethyl)oxy]ethanol, Dowanal EiPAT, Ethylene Glycol Isopropyl Ether, Ethylene Glycol Monisopropyl Ether, Ethylene Glycol Monoisopropyl Ether, Isopropyl Cellosolve, Isopropyl Oxitol, NSC 1259, Ucar AC, beta-Hydroxyethyl Isopropyl Ether
IUPAC Name 2-propan-2-yloxyethanol
SMILES CC(C)OCCO

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C5H12O2/c1-5(2)7-4-3-6/h5-6H,3-4H2,1-2H3
Formula Weight 104.0837
Chemical Name or Material 2-Isopropoxyethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.