Learn More
2-Iminobiotin, MedChemExpress
Product Code. 16447647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447647 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16447647 Supplier Medchem Express Supplier No. HY118700
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress 2-Iminobiotin (Guanidinobiotin) is a biotin (vitamin H or B7) analog. 2-Iminobiotin is a reversible nitric oxide synthases inhibitor with Kis of 21.8 and 37.5μM for murine iNOS and rat n-cNOS, respectively. 2-Iminobiotin superimposes on hypothermia protects human neuronal cells from hypoxia-induced cell damage.
2-Iminobiotin (Guanidinobiotin) is a biotin (vitamin H or B7) analog. 2-Iminobiotin is a reversible nitric oxide synthases inhibitor with Kis of 21.8 and 37.5μM for murine iNOS and rat n-cNOS, respectively. 2-Iminobiotin superimposes on hypothermia protects human neuronal cells from hypoxia-induced cell damage.
Specifications
Chemical Name or Material 2-Iminobiotin
CAS 13395-35-2
Color White
Purity Grade Notes Research
Molecular Formula C10H17N3O2S
Quantity 5 mg
Synonym Guanidinobiotin
Solubility Information DMSO : 12.5 mg/mL (51.37 mM; Need ultrasonic)
SMILES [H][C@](CS[C@H]1CCCCC(O)=O)([C@]1([H])N2)NC2=N
Molecular Weight (g/mol) 243.33
Formula Weight 243.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation