Learn More
2-(Hydroxymethyl)pyridine N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Hydroxymethyl)pyridine N-Oxide |
| CAS | 10242-36-1 |
| Molecular Formula | C6H7NO2 |
| Quantity | 50 mg |
| Synonym | 2-(Hydroxymethyl)pyridine 1-Oxide, 2-Pyridinemethanol 1-Oxide, 2-Pyridylcarbinol N-Oxide, 2-Pyridylmethanol N-Oxide, 2-Picolyl Alcohol 1-Oxide |
| InChI Formula | InChI=1S/C6H7NO2/c8-5-6-3-1-2-4-7(6)9/h1-4,8H,5H2 |
| SMILES | OCc1cccc[n+]1[O-] |
| IUPAC Name | (1-oxidopyridin-1-ium-2-yl)methanol |
| Molecular Weight (g/mol) | 125.13 |
| Formula Weight | 125.0477 |