missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzeneethanol, TRC

Product Code. 30394246
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394246 1 g -
30393442 5 g 5g
30389670 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394246 Supplier Toronto Research Chemicals Supplier No. TRCH8289851G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7768-28-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.16
Synonym 1,2-Tyrosol, 2-(2-Hydroxyethyl)phenol, 2-(2-Hydroxyphenyl)ethanol, 2-(2-Hydroxyphenyl)ethyl alcohol, 2-(o-Hydroxyphenyl)ethanol, 2-Hydroxybenzeneethanol, 2-Hydroxyphenethyl alcohol, NSC 101845, o-(2-Hydroxyethyl)phenol, o-Hydroxyphenethyl alcohol, beta-(o-Hydroxyphenyl)ethanol
IUPAC Name 2-(2-hydroxyethyl)phenol
SMILES OCCc1ccccc1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H10O2/c9-6-5-7-3-1-2-4-8(7)10/h1-4,9-10H,5-6H2
Formula Weight 138.0681
Chemical Name or Material 2-Hydroxybenzeneethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.