missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Fluorene, TRC

Product Code. 30012505
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012505 5 mg 5mg
30006454 50 mg 50mg
30003360 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012505 Supplier Toronto Research Chemicals Supplier No. TRCH9424305MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2443-58-5
Molecular Formula C13 H10 O
Molecular Weight (g/mol) 182.22
Synonym 9H-Fluoren-2-ol, Fluoren-2-ol (6CI,7CI,8CI), 2-Hydroxy-9H-fluorene, 2-Hydroxyfluorene, NSC 26316, NSC 31248
IUPAC Name 9H-fluoren-2-ol
SMILES Oc1ccc2c(Cc3ccccc23)c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H10O/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8,14H,7H2
Formula Weight 182.0732
Percent Purity >95%
Chemical Name or Material 2-Hydroxy Fluorene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.