Learn More
2-Hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic Acid |
| CAS | 101949-49-9 |
| Molecular Formula | C16H16O4 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C16H16O4/c1-16(2,10-3-6-12(17)7-4-10)11-5-8-14(18)13(9-11)15(19)20/h3-9,17-18H,1-2H3,(H,19,20) |
| SMILES | CC(C)(c1ccc(O)cc1)c2ccc(O)c(c2)C(=O)O |
| IUPAC Name | 2-hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic acid |
| Molecular Weight (g/mol) | 272.3 |
| Formula Weight | 272.1049 |