Learn More
2-Hydroxy-4-methylbenzenesulfonic Acid Ammonium Salt, TRC
Product Code. 30180308
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180308 250 mg 250mg
30180183 100 mg 100mg
30180005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30180308 Supplier Toronto Research Chemicals Supplier No. TRCH947480250MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79093-71-3
Molecular Formula C7 H8 O4 S . N H3
Molecular Weight (g/mol) 205.23
Synonym 2-Hydroxy-4-methylbenzenesulfonic Acid Ammonium Salt (1:1), Benzenesulfonic acid, 2-hydroxy-4-methyl-, monoammonium salt (9CI)
IUPAC Name azane;2-hydroxy-4-methylbenzenesulfonic acid
SMILES N.Cc1ccc(c(O)c1)S(=O)(=O)O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C7H8O4S.H3N/c1-5-2-3-7(6(8)4-5)12(9,10)11;/h2-4,8H,1H3,(H,9,10,11);1H3
Formula Weight 205.0409
Chemical Name or Material 2-Hydroxy-4-methylbenzenesulfonic Acid Ammonium Salt