missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methyl-3-nitropyridine, 98%, Thermo Scientific™

Product Code. 10383662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10383662 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10383662 Supplier Thermo Scientific Acros Supplier No. 319580050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21901-18-8
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H6N2O3
MDL Number MFCD00010689,MFCD00955770
Quantity 5 g
Synonym 2-hydroxy-4-methyl-3-nitropyridine, 4-methyl-3-nitropyridin-2-ol, 2-hydroxy-3-nitro-4-picoline, 2 1h-pyridinone, 4-methyl-3-nitro, 2-hydroxy-3-nitro-4-methylpyridine, 4-methyl-3-nitro-2-pyridone, 4-methyl-3-nitro-2-pyridinone, 4-methyl-3-nitro-2-pyridinol, 4-methyl-3-nitropyridin-2 1h-one, 4-methyl-3-nitro-1,2-dihydropyridin-2-one
InChI Key HZCWTTHQRMHIIE-UHFFFAOYSA-N
SMILES CC1=C(C(=O)NC=C1)[N+]([O-])=O
IUPAC Name 4-methyl-3-nitro-1H-pyridin-2-one
Molecular Weight (g/mol) 154.13
PubChem CID 345370
Formula Weight 154.13
Chemical Name or Material 2-Hydroxy-4-methyl-3-nitropyridine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.