missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methoxyacetophenone, 99%

Product Code. p-7026259
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11419707

Brand: Thermo Scientific Alfa Aesar B22173.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-41-0 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-41-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008730
InChI Key UILPJVPSNHJFIK-UHFFFAOYSA-N
Synonym paeonol, 2'-hydroxy-4'-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethanone, peonol, 2-hydroxy-4-methoxyacetophenone, 1-2-hydroxy-4-methoxyphenyl ethan-1-one, resacetophenone-4-methyl ether, ethanone, 1-2-hydroxy-4-methoxyphenyl, paeonal, paeonolum
PubChem CID 11092
ChEBI CHEBI:69581
IUPAC Name 1-(2-hydroxy-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)O

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1282794
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 2'-Hydroxy-4'-methoxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.