accessibility menu, dialog, popup

UserName

CAS RN 552-41-0

IUPAC Name: 1-(2-hydroxy-4-methoxyphenyl)ethan-1-one
Synonyms: paeonol 丹皮酚 4-O-methylresacetophenone peonol resacetophenone-4-methyl ether
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: UILPJVPSNHJFIK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=O)C(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)