Learn More
Alfa Aesar™ 2-Hydroxy-4,6-dimethylpyrimidine, 97%
Product Code. 11480284
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480284 100 g 100g
11470284 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11480284 Supplier Alfa Aesar™ Supplier No. A13676.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 108-79-2
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD00006072
InChI Key WHEQVHAIRSPYDK-UHFFFAOYSA-N
Synonym 2-hydroxy-4,6-dimethylpyrimidine, 4,6-dimethylpyrimidin-2-ol, 4,6-dimethyl-2-hydroxypyrimidine, 4,6-dimethylpyrimidin-2 1h-one, 2 1h-pyrimidinone, 4,6-dimethyl, unii-hg0cbn4v9f, 4,6-dimethyl-2-pyrimidinol, 4,6-dimethyl-pyrimidin-2-ol, hg0cbn4v9f, 4,6-dimethyl-2-pyrimidone
PubChem CID 9508
IUPAC Name 4,6-dimethyl-1H-pyrimidin-2-one
SMILES CC1=CC(C)=NC(=O)N1
Specifications
Melting Point 202°C to 204°C
Quantity 100 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4,6-dimethylpyrimidine