Learn More
(2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate, Thermo Scientific Chemicals
Catalog Number 15457705
Click to view available options
Quantity:
250 mg
1 g
896437-63-1
MFCD00078139
Quaternary ammonium beta-cylodextrin; QA-beta-CD
Questo articolo non è restituibile. Consulta la politica di reso
896437-63-1
MFCD00078139
Quaternary ammonium beta-cylodextrin; QA-beta-CD
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point ∽280°C (decomposition)
Quantity 250 mg
Chemical Name or Material (2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate