Learn More
2-Heptanone-1,1,1,3,3-d5, CDN
Product Code. 30319614
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319614 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319614 Supplier CDN Supplier No. CDND25161G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Heptanone-1,1,1,3,3-D5
CAS 24588-56-5
Molecular Formula C7 D5 H9 O
Quantity 1 g
Synonym 2-Heptanone-1,1,1,3,3-d5 (8 CI, 9 CI, ACI), 2-Heptanone-1,1,1,3,3-D5, 2-Heptanone-d5, Methyl-d3 pentyl-1,1-d2 ketone
InChI Formula InChI=1 S/C7H14O/c1-3-4-5-6-7(2)8/h3-6H2,1-2H3/i2D3,6D2
SMILES [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])CCCC
IUPAC Name 1,1,1,3,3-pentadeuterioheptan-2-one
Molecular Weight (g/mol) 119.2163
Formula Weight 119.1358 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity