missing translation for 'onlineSavingsMsg'
Learn More

2-Furoylglycine, TRC

Product Code. 30137986
Change view
Click to view available options
Quantity:
50 mg
100 mg
2.5 g
Unit Size:
100mg
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137986 100 mg 100mg
30135582 2.5 g 2.5g
30009518 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137986 Supplier Toronto Research Chemicals Supplier No. TRCF865225100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5657-19-2
Molecular Formula C7 H7 N O4
Molecular Weight (g/mol) 169.13
Synonym Glycine, N-2-furoyl- (8CI), N-(2-Furanylcarbonyl)glycine, 2-Furancarboxamide, N-(carboxymethyl)-, 2-Furoylglycine, 2-[(Furan-2-carbonyl)amino]acetic acid, Furfuroylglycine, Furoylglycine, N-(2-Furoyl)glycine, Pyromucuric acid, [(Furan-2-carbonyl)amino]acetic acid, [[(Furan-2-yl)carbonyl]amino]acetic acid
IUPAC Name 2-(furan-2-carbonylamino)acetic acid
SMILES OC(=O)CNC(=O)c1occc1

Specifications

InChI Formula InChI=1S/C7H7NO4/c9-6(10)4-8-7(11)5-2-1-3-12-5/h1-3H,4H2,(H,8,11)(H,9,10)
Quantity 100 mg
Formula Weight 169.0375
Shipping Condition Room Temperature
Chemical Name or Material Furoylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.