CAS RN 5657-19-2
CAS RN 5657-19-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Furoylglycine, TRC
CAS: 5657-19-2 Molecular Formula: C7 H7 N O4 Molecular Weight (g/mol): 169.13 Synonym: Glycine, N-2-furoyl- (8CI),N-(2-Furanylcarbonyl)glycine,2-Furancarboxamide, N-(carboxymethyl)-,2-Furoylglycine,2-[(Furan-2-carbonyl)amino]acetic acid,Furfuroylglycine,Furoylglycine,N-(2-Furoyl)glycine,Pyromucuric acid,[(Furan-2-carbonyl)amino]acetic acid,[[(Furan-2-yl)carbonyl]amino]acetic acid IUPAC Name: 2-(furan-2-carbonylamino)acetic acid SMILES: OC(=O)CNC(=O)c1occc1