missing translation for 'onlineSavingsMsg'
Learn More

2-Furanyl-1-piperazinylmethanone, TRC

Product Code. 30402769
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402769 10 g
30394292 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402769 Supplier Toronto Research Chemicals Supplier No. TRCF86404010G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40172-95-0
Molecular Formula C9 H12 N2 O2
Molecular Weight (g/mol) 180.2
Synonym 1-(Furan-2-ylcarbonyl)piperazine, Terazosin Hydrochloride Dihydrate Imp. L (EP), Prazosin Hydrochloride Imp. D (EP), Methanone, 2-furanyl-1-piperazinyl-, Piperazine, 1-(2-furanylcarbonyl)- (9CI), 2-Furanyl-1-piperazinylmethanone, (Furan-2-yl)(piperazin-1-yl)methanone, 1-(2-Furanoyl)piperazine, 1-(2-Furanylcarbonyl)piperazine, 1-(2-Furoyl)piperazine, 1-(2-Furylcarbonyl)piperazine, 1-(Furan-2-carbonyl)piperazine, 4-(Furan-2-ylcarbonyl)piperazine, N-(2-Furancarbonyl)piperazine, N-(2-Furoyl)piperazine, N-(Furan-2-oyl)piperazine
IUPAC Name furan-2-yl(piperazin-1-yl)methanone
SMILES O=C(N1CCNCC1)c2occc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H12N2O2/c12-9(8-2-1-7-13-8)11-5-3-10-4-6-11/h1-2,7,10H,3-6H2
Formula Weight 180.0899
Chemical Name or Material 1-(furan-2-ylcarbonyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.