Learn More
2-Fluoroethyl Chloroformate, TRC
Product Code. 30406970
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406970 10 g
30403967 1 g
30393605 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30406970 Supplier Toronto Research Chemicals Supplier No. TRCF59153010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 462-27-1
Molecular Formula C3H4ClFO2
Molecular Weight (g/mol) 126.51
Synonym Carbonochloridic Acid 2-Fluoroethyl Ester, 2-Fluoroethyl Carbonochloridate
SMILES FCCOC(=O)Cl
Specifications
Quantity 10 g
InChI Formula InChI=1S/C3H4ClFO2/c4-3(6)7-2-1-5/h1-2H2
Chemical Name or Material 2-Fluoroethyl Chloroformate